主要文件
CAD6002
3-way stopcock
female Luer lock to female Luer lock, right side, female Luer lock
登录查看公司和协议定价
选择尺寸
1 EA
$233.00
$233.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$233.00
About This Item
UNSPSC代码:
41122000
NACRES:
NB.03
$233.00
请联系客服了解存货情况
包装
pack of 1 ea
制造商/商品名称
Cadence Science 6002
正在寻找类似产品? 访问 产品对比指南
一般描述
The spring clip design holdsthe key in position under proper sealing pressure for optimum performance. There is no need for a screwdriver in cleaning operations.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi
历史批次信息供参考:
分析证书(COA)