主要文件
BR88680
BRAND®有嘴旋塞T孔
for tube I.D. 9 mm
别名:
PTFE, 旋塞
登录查看公司和协议定价
选择尺寸
1 EA
$121.00
$121.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$121.00
About This Item
UNSPSC代码:
41100000
NACRES:
NB.35
$121.00
请联系客服了解存货情况
物料
PTFE
包装
pack of 1 ea
制造商/商品名称
BRAND 88680
内径
5 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
法律信息
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)