Merck
BR4600230
BRAND®Dispensette® S 固定容量瓶口分液器
volume 5 mL, without recirculation valve
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121500
NACRES:
NB.01
特點
DE-M marking
without recirculation valve
製造商/商標名
BRAND 4600230
容積
5 mL
接頭
(GL 28/S28)
GL 32
GL 38
S 40
正在寻找类似产品? 访问 产品对比指南
一般說明
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
特點和優勢
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
其他說明
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
法律資訊
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
访问文档库
相关内容
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.