主要文件
BR43308
BRAND® BLAUBRAND®密度瓶,Gay-Lussac刻度,已校准
capacity 10 mL, borosilicate glass
别名:
BLAUBRAND®, Gay-Lussac
登录查看公司和协议定价
选择尺寸
1 EA
$76.70
$76.70
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$76.70
About This Item
UNSPSC代码:
41121800
NACRES:
NB.71
$76.70
请联系客服了解存货情况
物料
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
接合处
joint ST/NS neck10/19
特点
with cap
包装
pack of 1 ea
制造商/商品名称
BRAND 43308
容量
10 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
DIN ISO 3507, type Gay-Lussac. Calibrated to contain (TC, In). Incl. individual certificate. Stopper NS 10/19 with capillary. Top of stopper ground and polished. The volume in cm³ is specified to a precision of 3 decimal places.
法律信息
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
历史批次信息供参考:
分析证书(COA)
访问文档库
其他客户在看
实验方案
Density bottles determine liquid density, calibrated 'to contain' for accuracy, though not volumetric instruments like volumetric flasks.