主要文件
ACE729805
Ace addition funnel
capacity 125 mL
登录查看公司和协议定价
选择尺寸
1 EA
$584.00
$584.00
请联系客服了解存货情况
选择尺寸
变更视图
1 EA
$584.00
About This Item
UNSPSC代码:
41106514
NACRES:
NB.43
$584.00
请联系客服了解存货情况
特点
0-3 pressure equalizing funnel
容量
125 mL
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
Angled position makes manipulation of stopcock easier than conventional style. Leak-proof with complete drainage, pressure-equalizing arm, precise needle-valve adjustment, and double-scale graduations.
Threaded stopcock (0 to 3 mm orifice for funnels up to 250 mL and 0 to 5 mm orifice for funnels 500 mL and larger) with PTFE plug permits smooth needle-valve adjustment down to 0.1 mL/min flow rate. Double PTFE-ring seals prevent exposure of backup O-ring to corrosive liquids.
历史批次信息供参考:
分析证书(COA)