Skip to Content
Merck
All Photos(1)

Documents

06-519-AF647

Sigma-Aldrich

Anti-phospho-CREB (Ser133) Antibody, Alexa Fluor647 conjugate

from rabbit, ALEXA FLUOR 647

Synonym(s):

Cyclic AMP-responsive element-binding protein 1 (pSer133), cAMP-responsive element-binding protein 1 (pSer133), CREB-1 (pSer133), CREB-A (pSer133), htCREB (pSer133), CREB-B (pSer119)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), guinea pig (based on 100% sequence homology), Xenopus (based on 100% sequence homology), canine (based on 100% sequence homology), chicken (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunofluorescence: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer133)

Gene Information

human ... CREB1(1385)

General description

Cyclic AMP-responsive element-binding protein 1 (UniProt P16220; also known as cAMP-responsive element-binding protein 1, CREB-1) is encoded by the CREB1 gene (Gene ID 1385) in human. CREB is a transcription factor that binds the cAMP response elements (CRE) and regulates the transcription of the downstream genes. CREB activity is regulation via phosphorylation by various kinases, including PKA, and Ca2+/calmodulin-dependent protein kinases. Genes known to be regulated by CREB include c-fos, BDNF, tyrosine hydroxylase, neuropeptides (such as somatostatin, enkephalin, VGF, and corticotropin-releasing hormone), as well as the circadian clock genes PER1 and PER2. CREB is critical for memory formation across a wide range of species.

Specificity

Recognizes phosphorylated CREB-A (pSer133), htCREB (pSer133), and CREB-B (pSer119) spliced isoforms reported by (UniProt P16220). May also recognize phosphorylated CREM and ATF-1 that contain homologous sequence to that of the immungen peptide.

Immunogen

Epitope: pSer133.
KLH-conjugated synthetic peptide corresponding to the amino acids including and encompassing phosphorylated Ser133 of CREB-A.

Application

Anti-phospho-CREB (Ser133) Antibody, Alexa Fluor647 conjugate is an antibody against phospho-CREB for use in Immunofluorescence.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
The unconjugated antibody (Cat. No. 06-519) is shown to be suitable also for Electrophoretic Mobility Shift Assay (EMSA), immunohistochemistry, and Western blotting applications.

Quality

Evaluated by Immunofluorescence in rat brain tissue.

Immunofluorescence Analysis: A 1:50 dilution of this antibody detected phospho-CREB (Ser133) in rat brain tissue.

Target description

~43 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service