Skip to Content
Merck
All Photos(1)

Documents

W366501

Sigma-Aldrich

2,2-Dimethyl-5-(1-methylpropen-1-yl)tetrahydrofuran

98%, FG

Synonym(s):

Ocimene quintoxide, (±)-2,2-Dimethyl-5-(1-methyl-1-propenyl)tetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3665
Beilstein:
4797901
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.090

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

98%

refractive index

n20/D 1.4460 (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

citrus

SMILES string

C\C=C(/C)C1CCC(C)(C)O1

InChI

1S/C10H18O/c1-5-8(2)9-6-7-10(3,4)11-9/h5,9H,6-7H2,1-4H3/b8-5+

InChI key

LPEYLSKLVYWOEQ-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethyl-5-(1-methylpropen-1-yl)tetrahydrofuran is reported to be found in the essential oils of Artemisia species.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An investigation on the volatile composition of some Artemisia species from Iran.
Costa R, et al.
Flavour and Fragrance Journal, 24(2), 75-82 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service