Skip to Content
Merck
2,2-Dimethyl-5-(1-methylpropen-1-yl)tetrahydrofuran 98%, FG

2,2-Dimethyl-5-(1-methylpropen-1-yl)tetrahydrofuran

2,2-Dimethyl-5-(1-methylpropen-1-yl)tetrahydrofuran 98%, FG
Synonym(s):
Ocimene quintoxide, (±)-2,2-Dimethyl-5-(1-methyl-1-propenyl)tetrahydrofuran
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Brand
Biological Source
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing