Skip to Content
Merck
All Photos(1)

Documents

L3396

Sigma-Aldrich

Lead(II) acetate trihydrate

ACS reagent, 99.0-103.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(CH3CO2)2 · 3H2O
CAS Number:
Molecular Weight:
379.33
Beilstein:
3730298
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

ACS reagent

Assay

99.0-103.0%

form

solid

impurities

≤0.01% Insoluble matter

mp

75 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤5 ppm
nitrate and nitrite (as NO3-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.001%
K: ≤0.005%
Na: ≤0.01%

storage temp.

2-8°C

SMILES string

O.O.O.CC(=O)O[PbH2]OC(C)=O

InChI

1S/2C2H4O2.3H2O.Pb/c2*1-2(3)4;;;;/h2*1H3,(H,3,4);3*1H2;/q;;;;;+2/p-2

InChI key

MCEUZMYFCCOOQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirjam M Schaap et al.
Archives of toxicology, 89(12), 2413-2427 (2014-10-02)
Alternative methods to detect non-genotoxic carcinogens are urgently needed, as this class of carcinogens goes undetected in the current testing strategy for carcinogenicity under REACH. A complicating factor is that non-genotoxic carcinogens act through several distinctive modes of action, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service