17866
Streptavidin−CryptoFluor™ Crimson
suitable for fluorescence
About This Item
Recommended Products
mol wt
Mr ~40200 (αα′β2)(lit.)
fluorescence
λex 585 nm; λem ~658 nm in 0.1 M phosphate pH 7.2
suitability
suitable for fluorescence
storage temp.
2-8°C
General description
Application
Packaging
Reconstitution
Legal Information
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service