15025
meso-1,2-Bis(4-methoxyphenyl)ethylenediamine
for HPLC derivatization, LiChropur™, ≥98.0% (NT)
Synonym(s):
meso-1,2-Bis(4-methoxyphenyl)-1,2-diaminoethane, MOED
About This Item
Recommended Products
grade
for HPLC derivatization
Assay
≥98.0% (NT)
form
solid
quality
LiChropur™
mp
150-155 °C (lit.)
150-155 °C
fluorescence
λex 340 nm; λem 460 nm (Saccharide adducts)
application(s)
food and beverages
SMILES string
N[C@@H](C1=CC=C(OC)C=C1)[C@@H](C2=CC=C(OC)C=C2)N
InChI
1S/C16H20N2O2/c1-19-13-7-3-11(4-8-13)15(17)16(18)12-5-9-14(20-2)10-6-12/h3-10,15-16H,17-18H2,1-2H3/t15-,16+
InChI key
ZWMPRHYHRAUVGY-IYBDPMFKSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service