Merck
  • Isolation, purification and structure identification of two phenolic glycosides from the roots of Incarvillea younghusbandii Sprague and their antioxidant activities.
Isolation, purification and structure identification of two phenolic glycosides from the roots of Incarvillea younghusbandii Sprague and their antioxidant activities.
Yao xue xue bao = Acta pharmaceutica Sinica (2011-07-14)
Wei-gao Pan, Si-ping Jiang, Peng Luo, Ping Gao, Bin Chen, Hai-tao Bu
ABSTRACT
Using a bioassay-guided fractionation technique, two compounds were isolated from the roots of Incarvillea younghusbandii Sprague through silica gel, reverse-phase C18 column chromatography and reverse-phase HPLC. Their structures were identified as acteoside (1) and isoacteoside (2) by ESI-MS, GC-MS, 1D- and 2D-NMR. 1 and 2 showed *OH scavenging capacity similar with benzoic acid, higher O2*- (or *OH) scavenging capacity than ascorbic acid, far higher hepatic LPO inhibitory activities than 2, 6-di-tert-butyl-4-methylphenol (BHT) or ascorbic acid, and more powerful effect on protecting erythrocytes from oxidative damage than ascorbic acid. The *OH scavenging capacity was positively proportional to the concentrations of 1 and 2 ranging from 0.015 6 to 0.500 0 mg x mL(-1). The hepatic LPO inhibitory activities increased with the increasing concentrations of 1 and 2 from 0.001 9 to 0.250 0 mg x mL(-1), but decreased slightly with the increasing concentration from 0.250 0 to 1.0000 mg x L(-1).
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Verbascoside, primary reference standard