Skip to Content
Merck
All Photos(3)

Key Documents

V900365

Sigma-Aldrich

Octyl β-D-glucopyranoside

Vetec, reagent grade, 98%

Synonym(s):

n-Octyl glucoside, OGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Beilstein:
84118
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

reagent grade

description

non-ionic

product line

Vetec

Assay

98%

mol wt

micellar avg mol wt 25,000

aggregation number

84

CMC

20-25 mM (20-25°C)

transition temp

cloud point >100 °C

storage temp.

−20°C

SMILES string

CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic, dialyzable detergent for the solubilization and isolation of membrane proteins. Has been shown to increase the resolution of proteins in 2D gels.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Ménager et al.
Langmuir : the ACS journal of surfaces and colloids, 26(19), 15453-15463 (2010-09-10)
The present study deals with the morphological modifications of giant dioleoyl phosphatidylcholine vesicles (DOPC GUVs) induced by the nonionic surfactant n-octyl β,D-glucopyranoside at sublytic levels, i.e., in the first steps of the vesicle-to-micelle transition process, when surfactant inserts into the
Chaoqun Yao et al.
Proteomics. Clinical applications, 4(1), 4-16 (2010-12-08)
About two million new cases of leishmaniasis with 50 000 associated deaths occur worldwide each year. Promastigotes of the causative Leishmania spp. develop from the procyclic stage to the highly virulent metacyclic stage within the sand fly vector. We hypothesized
Zahrabatoul Mosapour Kotena et al.
Journal of molecular modeling, 19(2), 589-599 (2012-09-14)
Density functional theory calculations on two glycosides, namely, n-octyl-β-D-glucopyranoside (C(8)O-β-Glc) and n-octyl-β-D-galactopyranoside (C(8)O-β-Gal) were performed for geometry optimization at the B3LYP/6-31G level. Both molecules are stereoisomers (epimers) differing only in the orientation of the hydroxyl group at the C4 position.
Kerry K Karukstis et al.
The journal of physical chemistry. B, 116(12), 3816-3822 (2012-03-06)
When mixed with water, n-octyl β-D-glucoside forms self-assembled nanostructures, several of which are liquid crystalline and all of which depend on the water/glucoside ratio and temperature. For practical use of these phases, a detailed understanding of the conditions under which
Matthew J Ranaghan et al.
The journal of physical chemistry. B, 114(44), 14064-14070 (2010-10-23)
The photochemical and thermal stability of the detergent-solubilized blue- and green-absorbing proteorhodpsins, BPR and GPR, respectively, are investigated to determine the viability of these proteins for photonic device applications. Photochemical stability is studied by using pulsed laser excitation and differential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service