Skip to Content
Merck
All Photos(1)

Key Documents

11140922001

Roche

CTP

Lithium salt

Synonym(s):

lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

Assay

≥98% (HPLC)

form

solution

packaging

pkg of 400 μL (40 μmol; 100mM)

manufacturer/tradename

Roche

shipped in

dry ice

storage temp.

−20°C

General description

CTP is a 100 mM solution of the lithium salt (pH 7).

Application

CTP, lithium salt, is suitable for applications such as in vitro RNA transcription.

Quality

Typical analysis:
  • ≥98% CTP (HPLC), ≤1.5% CDP (HPLC), ≤0.5% CMP (HPLC)
  • Tested for the absence of RNases according to the current Quality Control procedures.

Specifications

Absorbance: 1mM CTP = 9.6 A272 units

Unit Definition

Unit Definition: 1 mM CTP = 9.6 A272 units

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidized low-density lipoprotein is a novel predictor of interferon responsiveness in chronic hepatitis C infection
Solbach, P, et al.
Cellular and molecular gastroenterology and hepatology, 1.3, 285-294 (2015)
Riku Nagai et al.
Methods and protocols, 4(3) (2021-07-22)
The rates of translation elongation or termination in eukaryotes are modulated through cooperative molecular interactions involving mRNA, the ribosome, aminoacyl- and nascent polypeptidyl-tRNAs, and translation factors. To investigate the molecular mechanisms underlying these processes, we developed an in vitro translation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service