Skip to Content
Merck
All Photos(3)

Key Documents

MAK558

Sigma-Aldrich

Trypsin Activity Assay Kit

sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104

usage

sufficient for 100 colorimetric tests

Quality Level

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

storage temp.

−20°C

General description

Trypsin is a pancreatic serine protease, which hydrolyzes peptide bonds specifically at the carboxyl side of arginine and lysine residues. The rate of hydrolysis is slower if an acidic residue is on either side of the cleavage site and cleavage may not occur if a proline residue is on the carboxyl side. Tryptic digestion of the protein of interest results in a highly specific cleavage and a limited number of peptide fragments. In this assay, trypsin cleaves a substrate to generate p-nitroaniline (p-NA) which is detected at λ= 405 nm. Trypsin activity can be accurately measured by the color intensity which is proportional to p-NA content. The kit detects 10–100 mU (p-NA unit) trypsin in various samples.

Features and Benefits

Highly sensitive, simple and high-throughput assay for measuring Trypsin concentration.
Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.

Click here to download a calculator excel file

Suitability

Suitable for the detection of Trypsin in various biological samples such as cell and tissue extracts, serum, and plasma.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service