Skip to Content
Merck
All Photos(1)

Key Documents

NIST90

Ferrophosphorus

NIST® SRM® 90

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 75 g

manufacturer/tradename

NIST®

format

matrix material

storage temp.

15-25°C

General description

This Standard Reference Material (SRM) is ferrophosphorus, an alloy used in manufacturing steel. SRM 90 is intended for use in calibration and the evaluation of chemical and instrumental methods of analysis. A unit of SRM 90 consists of a bottle containing approximately 75 g of powder. Certified Values: The certified value for phosphorus in SRM 90 is reported in Table 1. Please refer to the SDS and COA for more information.

SRM 90_cert

SRM 90_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Phosphorus (P)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service