Skip to Content
Merck
All Photos(1)

Key Documents

1.46711

Millipore

Polymyxin Pyruvate Egg Yolk Mannitol Bromothymol Blue Agar

ReadyPlate 90mm plate in accordance with ISO for the enumeration of Bacillus cereus in food samples

Synonym(s):

PEMB Agar, Bacillus Cereus Agar

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€260.00

€260.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
100 EA
€260.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€260.00


Please contact Customer Service for Availability

Request more information

Agency

ISO 21871

Quality Level

form

plate (ready-to-use)

shelf life

4 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (90 mm plate (100 plates per pack))

pH

7.2 ( in H2O)

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Bacillus cereus

General description

ReadyPlate agar plates are stackable, specially designed plates that help increase safety during handling, transport, and incubation. ReadyPlate agar plates are of standard 90mm diameter. PEMBA is a highly specific, selective medium for the isolation and enumeration of B. cereus from foods. Polymyxin and a critical concentration of nutrients make this media selective. It supports the growth of even a small number of B. cereus cells and spores from foods having a high microbial load. The low peptone content in the medium promotes sporulation and sodium pyruvate reduces the colony size of the organisms. Egg yolk emulsion demonstrates the strong lecithinase opacity reaction. Bromothymol blue acts as a pH indicator to detect mannitol fermentation. The addition of polymyxin B sulfate makes the medium selective for the isolation of B. cereus. Cycloheximide inhibits the growth of molds.

Application

PEMBA is a highly specific, selective medium for the isolation and enumeration of B. cereus from food samples.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Interview: Accurate microbiology QC methods for infant formula testing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service