MilliporeSigma
HPLC Analysis of Bronchodilators Theophylline and Other Caffeine Metabolites in Serum on Discovery® RP-AmideC16 after SPE using Discovery® DSC-18
HPLC Analysis of Bronchodilators Theophylline and Other Caffeine Metabolites in Serum on Discovery® RP-AmideC16 after SPE using Discovery® DSC-18 application for SPE, application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Discovery® RP-AmideC16 HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
SPE tube or plate
Product No.
Description
Pricing
Discovery® DSC-18 SPE Tube
bed wt. 500 mg, volume 3 mL, pk of 54
CONDITIONS
sample preparation
SPE (Solid Phase Extraction)
SPE tube/cartridge
Discovery® DSC-18, 500 mg/3 mL (52603-U)
condition
2 mL methanol, then 2 mL water
sample preparation
1 mL porcine serum spiked with 0.1 μg/mL, 0.50 μg/mL, or 1.0 μg/mL each analyte
sample preparation
1 mL methanol; evaporate to dryness with nitrogen stream at room temperature; reconstitute in 200 μL mobile phase containing 0.2 μg/mL or 7-methylxanthine (IS)
sample preparation
2 mL 5% methanol in water; dry tube 10 min with nitrogen stream
column
Discovery RP-AmideC16, 15 cm × 4.6 mm, 5 μm particles, preceded by a 2 cm RP-AmideC16 guard column and 0.5 μm frit filter (505013)
mobile phase
[A] Methanol; [B] 1% acetic acid (17:38, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 272 nm
injection
20 μL reconstituted porcine serum extract
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany