MilliporeSigma
  • Aurora kinase B dependent phosphorylation of 53BP1 is required for resolving merotelic kinetochore-microtubule attachment errors during mitosis.
Aurora kinase B dependent phosphorylation of 53BP1 is required for resolving merotelic kinetochore-microtubule attachment errors during mitosis.
Oncotarget (2017-04-19)
Haibo Wang, Bin Peng, Raj K Pandita, David A Engler, Risë K Matsunami, Xingzhi Xu, Pavana M Hegde, Brian E Butler, Tej K Pandita, Sankar Mitra, Bo Xu, Muralidhar L Hegde
ABSTRACT
Defects in resolving kinetochore-microtubule attachment mistakes during mitosis is linked to chromosome instability associated with carcinogenesis as well as resistance to cancer therapy. Here we report for the first time that tumor suppressor p53-binding protein 1 (53BP1) is phosphorylated at serine 1342 (S1342) by Aurora kinase B both in vitro and in human cells, which is required for optimal recruitment of 53BP1 at kinetochores. Furthermore, 53BP1 staining normally localized on the outer kinetochore, extended to the whole kinetochore when it is merotelically-attached, in concert with mitotic centromere-associated kinesin. Kinetochore-binding of pS1342-53BP1 is essential for efficient resolving of merotelic attachment, a spontaneous kinetochore-microtubule connection error that usually causes aneuploidy. Consistently, loss of 53BP1 results in significant increase in lagging chromosome events, micronuclei formation and aneuploidy, due to the unresolved merotely in both cancer and primary cells, which is prevented by ectopic wild type 53BP1 but not by the nonphophorylable S1342A mutant. We thus document a novel DNA damage-independent function of 53BP1 in maintaining faithful chromosome segregation during mitosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF2C