- Corneal surface glycosylation is modulated by IL-1R and
Corneal surface glycosylation is modulated by IL-1R and
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-02-23)
Amber L Jolly, Paresh Agarwal, Matteo M E Metruccio, David R Spiciarich, David J Evans, Carolyn R Bertozzi, Suzanne M J Fleiszig
PMID28223334
ABSTRACT
Cell surface glycosylation is thought to be involved in barrier function against microbes at mucosal surfaces. Previously we showed that the epithelium of healthy mouse corneas becomes vulnerable to