MilliporeSigma
  • Maltol inhibits apoptosis of human neuroblastoma cells induced by hydrogen peroxide.
Maltol inhibits apoptosis of human neuroblastoma cells induced by hydrogen peroxide.
Journal of biochemistry and molecular biology (2006-04-06)
Yang Yang, Jian Wang, Caimin Xu, Huazhen Pan, Zinan Zhang
ABSTRACT
To analyze the effect of Maltol on the apoptosis of Human Neuroblastoma Cells (SH-SY5Y) treated by free radical which was generated from Hydrogen Peroxide (H2O2), flow cytometry analysis on Phosphatidylserine (PS) inverting percentage was applied to determine the apoptosis. MTT (3-(4,5-dimethythiazol-2-yl)-2,5-diphenyl-tetrazolium bromide) assay was employed to analyze the cell viability. DNA electrophoresis was used to detect DNA fragmentation. Moreover intracellular calcium of concentration ([Ca2+]i) was measured by fluorescence emission. Flow cytometry analysis on the function of mitochondria and Western blot analysis of NF-kappaB. The results showed that the pretreatment with maltol for 2 hours could prevent the H2O2-induced apoptosis. Maltol could reduce the inverting percentage of PS, DNA fragmentation and [Ca2+]i, and enhance the cellular function of mitochondria. NF-kappaB activated by H2O2 is reduced. The experiments suggest that maltol could effectively inhibit the apoptosis induced by H2O2. As a novel anti-oxidant, maltol is a new promising drug in protecting the neurological cells from the damage by free radical.
Sigma-Aldrich
C7BzO