MilliporeSigma
  • Quantitative confirmation of dimetridazole and ipronidazole in swine feed by capillary gas chromatography/mass spectrometry with multiple ion detection.
Quantitative confirmation of dimetridazole and ipronidazole in swine feed by capillary gas chromatography/mass spectrometry with multiple ion detection.
Journal - Association of Official Analytical Chemists (1987-07-01)
W J Morris, G J Nandrea, J E Roybal, R K Munns, W Shimoda, H R Skinner
ABSTRACT
Extracts from 4 types of swine feed containing 0.11 ppm each of dimetridazole (DMZ) and ipronidazole (IPR) were analyzed by capillary gas chromatography/mass spectrometry (GC/MS) using multiple ion detection (MID) techniques. We demonstrate in this paper that the quantitative results obtained by capillary GC/MS with MID are comparable for both compounds to results obtained by liquid chromatography and have a lower coefficient of variation for DMZ. Moreover, consistency in the ion ratios (5 ions in DMZ and 6 ions in IPR) permits identification of these compounds by electron ionization MS.
Product Description
Supelco
Ipronidazole, VETRANAL®, analytical standard
Pricing and availability is not currently available.