MilliporeSigma
  • Crystal structure, differential scanning calorimetric, infrared spectroscopy and theoretical studies of C(NH2)2(NH)*CH2=CHCOOH noncentrosymmetric crystal.
Crystal structure, differential scanning calorimetric, infrared spectroscopy and theoretical studies of C(NH2)2(NH)*CH2=CHCOOH noncentrosymmetric crystal.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-10)
M Drozd, D Dudzic, A Pietraszko
ABSTRACT
The X-ray and vibrational spectroscopic analysis of a new molecular complex of guanidine and acrylic acid are reported. The crystal of C(NH(2))(2)(NH)(*)CH(2)=CHCOOH belongs to Pna2(1) space group of orthorhombic system with Z=4, a=9.9242(34) Å, b, c=8.3951(14) Å. In the crystal structure the macroscopic symmetry center is absent. The differential scanning calorimetric (DSC) experiment of powder sample indicates on continuous phase transition at 235.8K. The room temperature infrared spectrum of guanidine*acrylic acid powder sample has been measured. The spectrum is discussed on the basis of crystallographic data. The IR spectra for the powder sample were also measured at low temperatures (12-300 K). The temperature relationships of band position for obtained spectra are analyzed. Additionally, the results of theoretical calculation of vibrational spectra, equilibrium structure, HOMO, LUMO and first order hyperpolarizability are presented.
Sigma-Aldrich
Zinc acrylate, 98%
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%