MilliporeSigma
  • Importance of the correct diagnosis of opioid-induced respiratory depression in adult cancer patients and titration of naloxone.
Importance of the correct diagnosis of opioid-induced respiratory depression in adult cancer patients and titration of naloxone.
Clinical medicine (London, England) (2013-05-18)
Jason Boland, Elaine Boland, David Brooks
ABSTRACT
Opioids can induce respiratory depression by invoking a centrally mediated decrease in involuntary respiratory rate, which in severe cases can cause a decrease in oxygen saturation. If respiratory depression is opioid induced, both low respiratory rate and low oxygen saturation will be present. If this is the case, oxygenation, rousing by verbal and physical stimulation and decreasing the opioid dose should be tried first. Naloxone, an opioid antagonist, should be avoided if at all possible but, if essential, titrate slowly to respiratory function administering 20-100 µg intravenously every two minutes. If used as a bolus for a patient on long-term opioids for chronic cancer pain, then refractory pain and symptomatic opioid withdrawal can result.
Sigma-Aldrich
Naloxone hydrochloride dihydrate, ≥98% (TLC and titration), powder
Pricing and availability is not currently available.
Supelco
Naloxone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.