MilliporeSigma
  • Synthesis and redox behavior of biferrocenyl-functionalized ruthenium(II) terpyridine gold clusters.
Synthesis and redox behavior of biferrocenyl-functionalized ruthenium(II) terpyridine gold clusters.
Langmuir : the ACS journal of surfaces and colloids (2004-10-06)
Teng-Yuan Dong, Hao-Wei Shih, Ling-Shao Chang
ABSTRACT
Spectroscopic and electrochemical characterizations of ferrocene- and biferrocene-functionalized terpyridine octanethiolate monolayer-protected clusters were investigated and reported. The electrochemical measurements of Ru2+ coordinated with 4'-ferrocenyl-2,2':6',2' '-terpyridine and 4'-biferrocenyl-2,2':6',2' '-terpyridine complexes were dominated by the Ru2+/Ru3+ redox couple (E(1/2) at approximately 1.3 V), Fe(2+)/Fe(3+) redox couples (E(1/2) from approximately 0.6 to approximately 0.9 V), and terpy/terpy-/terpy2- redox couples (E(1/)(2) at ca. -1.2 and ca. -1.4 V). The substantial appreciable variations detected in the Ru2+/Ru3+ and Fe2+/Fe3+ oxidation potentials indicate that there is an interaction between the Ru2+ and Fe2+ metal centers. The coordination of the Ru2+ metal center with 4'-ferrocenyl-2,2':6',2' '-terpyridine and 4'-biferrocenyl-2,2':6',2' '-terpyridine leads to an intense 1[(d(pi)Fe)6] --> 1[d(pi)Fe)5(pi*terpyRu)1] transition in the visible region. The 1[(d(pi)Fe)6] -->1[d(pi)Fe)5(pi*terpyRu)1] transition observed at approximately 510 nm revealed that there was a qualitative electronic coupling between metal centers. The coordination of the Ru2+ transition metal center lowers the energy of the pi*terpy orbitals, causing this transition.
Sigma-Aldrich
2,2′:6′,2′′-Terpyridine, 98%