MilliporeSigma
  • Switching gears: translational mastery of transcription during memory formation.
Switching gears: translational mastery of transcription during memory formation.
Neuron (2007-04-20)
Charles A Hoeffer, Eric Klann
ABSTRACT
A critical molecular requirement underlying many forms of long-lasting synaptic plasticity and memory is the de novo synthesis of mRNAs and proteins. In a recent paper in Cell, Costa-Mattioli et al. present data from a pharmacogenetic study that places a key regulatory event in the "neural decision" to undergo these persistent neuronal changes under translational control mediated by eIF2alpha.
Sigma-Aldrich
Sal003, ≥98% (HPLC)
Pricing and availability is not currently available.