MilliporeSigma
  • Brain-Implantable Multifunctional Probe for Simultaneous Detection of Glutamate and GABA Neurotransmitters.
Brain-Implantable Multifunctional Probe for Simultaneous Detection of Glutamate and GABA Neurotransmitters.
Sensors and actuators. B, Chemical (2022-05-24)
Nicolaie Moldovan, Iuliu-Ioan Blaga, Sanjeev Billa, Imran Hossain, Chenggong Gong, Claire E Jones, Teresa A Murray, Ralu Divan, Shabnam Siddiqui, Prabhu U Arumugam
ABSTRACT
Glutamate (GLU) and gamma-aminobutyric acid (GABA) are neurotransmitters (NTs) with an essential role in signal transmission in the brain. Brain disorders, such as epilepsy, Alzheimer's and Parkinson's diseases, and traumatic brain injury can be linked to imbalances in the GLU-GABA homeostasis that occurs in sub-second to seconds time frames. Current measurement techniques can detect these two NT concentrations simultaneously only in vitro. The present work reports on the fabrication of a silicon multifunctional biosensor microarray probe for sub-second simultaneous GLU-GABA detection in real-time, with excellent analyte sensitivity and selectivity and in vivo capabilities. The novel Si probes feature four surface-functionalized platinum ultramicroelectrodes (UMEs) for simultaneous amperometric detection of GLU and GABA with a sentinel, and a built-in microfluidic channel for the introduction of neurochemicals in the proximity of the UMEs. The microchannel also allows functioning of an On-Demand In-situ Calibrator that runs in-situ biosensor calibration. The probe exhibited excellent robustness at insertion in agarose-gel brain-tissue-mimicking test, and remarkably high hydrogen peroxide sensitivity (a by-product of GLU-GABA enzyme biosensor) with values on the order of 5000 nA μM -1 cm -2 and maximum sensitivities of 204±15 nA μM -1 cm -2 and 37±7 nA μM -1 cm -2 for GLU and GABA, respectively. Furthermore, the limit of detection of the biosensors reached as low as 7 nM, 165 nM and 750 nM for H 2 O 2, GLU and GABA, respectively and a temporal resolution of hundreds of milliseconds during in vivo studies using freely moving rats.
Sigma-Aldrich
GABase from Pseudomonas fluorescens, lyophilized powder, Protein ≥30 % by biuret