MilliporeSigma
  • Dataset on optimization and development of a point-of-care glucometer-based SARS-CoV-2 detection assay using aptamers.
Dataset on optimization and development of a point-of-care glucometer-based SARS-CoV-2 detection assay using aptamers.
Data in brief (2021-08-18)
Naveen K Singh, Partha Ray, Aaron F Carlin, Sydney C Morgan, Celestine Magallanes, Louise C Laurent, Eliah S Aronoff-Spencer, Drew A Hall
ABSTRACT
We present supplementary data for the published article, "Hitting the diagnostic sweet spot: Point-of-care SARS-CoV-2 salivary antigen testing with an off-the-shelf glucometer" [1]. The assay described is designed to be performed at home or in a clinic without expensive instrumentation or professional training. SARS-CoV-2 is detected by an aptamer-based assay that targets the Nucleocapsid (N) or Spike (S) antigens. Binding of the N or S protein to their respective aptamer results in the competitive release of a complementary antisense-invertase enzyme complex. The released enzyme then catalyzes the conversion of sucrose to glucose that is measured by an off-the-shelf glucometer. The data presented here describe the optimization of the assay parameters and their contribution to developing this aptamer-based assay to detect SARS-CoV-2. The assay performance was checked in a standard buffer, contrived samples, and patient samples validated with well-established scientific methods. The resulting dataset can be used to further develop glucometer-based assays for diagnosing other communicable and non-communicable diseases.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Microcentrifuge tubes with attached lid, capacity 1.5 mL
Pricing and availability is not currently available.