MilliporeSigma
  • Resolvin-D2 targets myogenic cells and improves muscle regeneration in Duchenne muscular dystrophy.
Resolvin-D2 targets myogenic cells and improves muscle regeneration in Duchenne muscular dystrophy.
Nature communications (2021-10-31)
Junio Dort, Zakaria Orfi, Paul Fabre, Thomas Molina, Talita C Conte, Karine Greffard, Ornella Pellerito, Jean-François Bilodeau, Nicolas A Dumont
ABSTRACT
Lack of dystrophin causes muscle degeneration, which is exacerbated by chronic inflammation and reduced regenerative capacity of muscle stem cells in Duchenne Muscular Dystrophy (DMD). To date, glucocorticoids remain the gold standard for the treatment of DMD. These drugs are able to slow down the progression of the disease and increase lifespan by dampening the chronic and excessive inflammatory process; however, they also have numerous harmful side effects that hamper their therapeutic potential. Here, we investigated Resolvin-D2 as a new therapeutic alternative having the potential to target multiple key features contributing to the disease progression. Our in vitro findings showed that Resolvin-D2 promotes the switch of macrophages toward their anti-inflammatory phenotype and increases their secretion of pro-myogenic factors. Moreover, Resolvin-D2 directly targets myogenic cells and promotes their differentiation and the expansion of the pool of myogenic progenitor cells leading to increased myogenesis. These effects are ablated when the receptor Gpr18 is knocked-out, knocked-down, or blocked by the pharmacological antagonist O-1918. Using different mouse models of DMD, we showed that Resolvin-D2 targets both inflammation and myogenesis leading to enhanced muscle function compared to glucocorticoids. Overall, this preclinical study has identified a new therapeutic approach that is more potent than the gold-standard treatment for DMD.
Sigma-Aldrich
Anti-GPR18 antibody produced in rabbit, affinity isolated antibody