MilliporeSigma
  • First Insights on Early Host Plants and Dispersal Behavior of Halyomorpha halys (Hemiptera: Pentatomidae) from Overwintering to Crop Colonization.
First Insights on Early Host Plants and Dispersal Behavior of Halyomorpha halys (Hemiptera: Pentatomidae) from Overwintering to Crop Colonization.
Insects (2020-12-10)
Lara Bosco, Martina Nardelli, Luciana Tavella
ABSTRACT
Following its first detection in North Italy in 2012, H. halys has become a serious threat in many crops, including hazelnut. The present study aimed at investigating dispersal capacity and behavior in relation to host plants of overwintered adults of H. halys before the colonization of hazelnut crop. Research was carried out in four polyculture areas (from 14 to 50 ha) in north-western Italy in 2018, by using (i) pheromone-baited traps, (ii) visual inspection and beating sheet sampling, and (iii) immunomarking-capture technique. The relative abundance of H. halys was similar between and within the study areas, and the early attractiveness of lures to adults after overwintering was confirmed; the host plants near a pheromone trap (less than 5 m) hosted higher numbers of H. halys than the same plant species far away. Hybrid plane, European spindletree, walnut, oak, and European elder were the first plants on which adult bugs were observed to feed. By immunomarking-capture technique, H. halys showed both short- and long-range dispersal from overwintering sites and/or early host plants to wild and crop plants. Marked adults were found in all zones of each area, irrespective of the distance from the protein treatment. Therefore, H. halys movement patterns depend on the ecosystem features, and plant host distribution and availability. This knowledge together with the interaction between pheromone and early host plants might contribute to the management of post-overwintering adults.
Sigma-Aldrich
Anti-Chicken Egg Albumin antibody produced in rabbit, whole antiserum
Pricing and availability is not currently available.