MilliporeSigma
  • The ratiometric fluorescence nanoparticle based on SiRB for pH detection of tumor.
The ratiometric fluorescence nanoparticle based on SiRB for pH detection of tumor.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2018-03-20)
Qinghui Wang, Xiaoyu Ding, Yuhui Wang, Qian Du, Tianguo Xu, Bin Du, Hanchun Yao
ABSTRACT
Tumor pH detection and pH value change monitoring have been of great interest in the field of nanomedicine. In this study, a pH-sensitive near-infrared fluorescence probe SiRB (Si-rhodamine and Boronic acid group) was synthesized by introducing a boronic acid group into the silicon rhodamine structure. ICG (Indocyanine green) as the fluorescence internal standard and SiRB were loaded into PLGA (poly lactic-co-glycolic acid) to form PLGA-SiRB-ICG nanoparticle. The experiments showed that the size of the nanoparticle was about 90 nm, which can reach tumor passively by enhancing permeability and retention effect. PLGA in the acidic environment will accelerate the release of cleavage, and the fluorescence ratio of the two probes can reflect the specific pH value in the tumor. The results indicated that the nanoparticle could quantitatively measure the pH value of the tumor site, which is expected to be used in tumor research and treatment.
Sigma-Aldrich
sec-Butyllithium solution, 1.4 M in cyclohexane