Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z703575

Sigma-Aldrich

Electrophoresis in Practice: A Guide to Methods and Applications of DNA and Protein Separations, 4th ed.

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

ISBN-10:
3-52731181-5
ISBN-13:
978-3-52731181-1
UNSPSC Code:
55101509

publication info

R. Westermeier, John Wiley & Sons, 2005, 384 pp., hard cover

packaging

pkg of 1 ea

manufacturer/tradename

Wiley

General description

This laboratory guide for successful electrophoretic separations is divided into two parts to provide readers with a thorough presentation of the fundamentals followed by a detailed description of the most common methods currently in use. This fourth edition retains the successful concept of its predecessors, yet features a brand-new layout, and is further enhanced by a section on difference gel electrophoresis, while the chapter on proteome analysis is practically all new and considerably extended, plus there are now around 10 % new literature references.

Table of Contents

Part I Fundamentals.
1 Electrophoresis.
1.0 General.
1.1 Electrophoresis in non-restrictive gels.
1.2 Electrophoresis in restrictive gels.
2 Isotachophoresis.
2.1 Migration with the same speed.
2.2 Ion train separation.
2.3 Zone sharpening effect.
2.4 Concentration regulation effect.
3 Isoelectric focusing.
3.1 Principles.
3.2 Gels for IEF.
3.3 Temperature.
3.4 Controlling the pH gradient.
3.5 The kinds of pH gradients.
3.6 Protein detection in IEFgels.
3.7 Preparative isoelectric focusing.
3.8 Titration curve analysis.
4 Blotting.
4.1 Principle.
4.2 Transfer methods.
4.3 Blotting membranes.
4.4 Buffers for electrophoretic transfers.
4.5 General staining.
4.6 Blocking.
4.7 Specific detection.
4.8 Protein sequencing.
4.9 Transfer problems.
5 Interpretation of electropherograms.
5.1 Introduction.
5.2 Image analysis.
6 Proteome Analysis.
6.1 General.
6.2 Sample preparation.
6.3 Two-dimensional electrophoresis.
6.4 Detection techniques.
6.5 Image analysis.
6.6 Protein spot identification.
6.7 Bioinformatics.
6.8 Functional proteomics.
7 Instrumentation.
7.1 Current and voltage conditions.
7.2 Power supply.
7.3 Separation chambers.
7.4 Staining apparatus for gels and blots.
7.5 Automated electrophoresis.
7.6 Instruments for 2-D electrophoresis.
7.7 Safety measures.
7.8 Environmental aspects.
Part II Equipment and Methods.
Equipment for Part II.
Instrumentation.
Special laboratory equipment.
Consumables.
Chemicals.
Method 1: PAGE of dyes.
Method 2: Agarose and immuno electrophoresis.
Method 3: Titration curve analysis.
Method 4: Native PAGE in amphoteric buffers.
Method 5: Agarose IEF.
Method 6: PAGIEF in rehydrated gels.
Method 7: Horizontal SDS-PAGE.
Method 8: Vertical PAGE.
Method 9: Semi-dry blotting of proteins.
Method 10: IEF in immobilized pH gradients.
Method 11: High-resolution 2-D electrophoresis.
Method 12: PAGE of double stranded DNA.
Method 13: Native PAGE of single stranded DNA.
Method 14: Denaturing gradient gel electrophoresis.
Method 15: Denaturing PAGE of DNA.
Appendix Trouble-shooting.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service