47305
3-(Fmoc-amino)-1-propanol
≥98.0%
Synonym(s):
9-Fluorenylmethyl N-(3-hydroxypropyl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
assay
≥98.0%
form
solid
reaction suitability
reagent type: cross-linking reagent
functional group
Fmoc
hydroxyl
storage temp.
2-8°C
SMILES string
OCCCNC(OCC1C(C=CC=C2)=C2C3=C1C=CC=C3)=O
InChI
1S/C18H19NO3/c20-11-5-10-19-18(21)22-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17,20H,5,10-12H2,(H,19,21)
InChI key
GNXZNUJDAMSJFZ-UHFFFAOYSA-N
Other Notes
Building block for the synthesis of oxa-analogues and homologues of naturally occurring polyamines.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tetrahedron Letters, 35, 3605-3605 (1994)
Synthesis, 859-859 (1998)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service