443883
Lanthanum(III) perchlorate solution
40 wt. % in H2O, 99.9% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
La(ClO4)3
CAS Number:
Molecular Weight:
437.26
EC Number:
MDL number:
UNSPSC Code:
12352302
Recommended Products
vapor pressure
0.12 psi ( 20 °C)
1.68 psi ( 55 °C)
assay
99.9% trace metals basis
form
liquid
concentration
40 wt. % in H2O
density
1.479 g/mL at 25 °C
SMILES string
O.[La+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wolfram W Rudolph et al.
Dalton transactions (Cambridge, England : 2003), 44(1), 295-305 (2014-11-08)
Raman spectra of aqueous lanthanum perchlorate, triflate (trifluorosulfonate), chloride and nitrate solutions were measured over a broad concentration (0.121-3.050 mol L(-1)) range at room temperature (23 °C). A very weak mode at 343 cm(-1) with a full width at half
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service