- Combined SEP and anti-PD-L1 antibody produces a synergistic antitumor effect in B16-F10 melanoma-bearing mice.
Combined SEP and anti-PD-L1 antibody produces a synergistic antitumor effect in B16-F10 melanoma-bearing mice.
Scientific reports (2018-01-11)
Zhengping Hu, Liang Ye, Yingying Xing, Jinhang Hu, Tao Xi
PMID29317734
ABSTRAKT
The increased PD-L1 induces poorer prognosis in melanoma. The treatment with PD-1/PD-L1 antibodies have a low response rate. The combination immunotherapies are the encouraging drug development strategy to receive maximal therapeutic benefit. In this study, we investigated the enhanced antitumor and immunomodulatory activity of combined SEP and αPD-L1 in B16-F10 melanoma-bearing mice. The results shown that combined SEP and αPD-L1 presented significant synergistic antitumor effects, increased the frequency of CD8
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Bovine IL2 / Interleukin-2 ELISA Kit, for serum, plasma and cell culture supernatants