- Tissue-specific regulation of BMP signaling by
Tissue-specific regulation of BMP signaling by
eLife (2017-08-23)
Antonio Galeone, Seung Yeop Han, Chengcheng Huang, Akira Hosomi, Tadashi Suzuki, Hamed Jafar-Nejad
PMID28826503
ABSTRAKT
Mutations in the human
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-HA-Biotyna, mysie przeciwciało monoklonalne, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NGLY1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution