Merck
Rapid biodiesel synthesis from waste pepper seeds without lipid isolation step.
Bioresource technology (2017-05-14)
Jechan Lee, Jieun Kim, Yong Sik Ok, Eilhann E Kwon
ABSTRAKT
In situ transformation of lipid in waste pepper seeds into biodiesel (i.e., fatty acid methyl esters: FAMEs) via thermally-induced transmethylation on silica was mainly investigated in this study. This study reported that waste pepper seeds contained 26.9wt% of lipid and that 94.1% of the total lipid in waste pepper seeds could be converted into biodiesel without lipid extraction step for only ∼1min reaction time. This study also suggested that the optimal temperature for in situ transmethylation was identified as 390°C. Moreover, comparison of in situ process via the conventional transmethylation catalyzed by H
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL