Merck
MicroRNA-146a Promotes Oligodendrogenesis in Stroke.
Molecular neurobiology (2016-01-08)
Xian Shuang Liu, Michael Chopp, Wan Long Pan, Xin Li Wang, Bao Yan Fan, Yi Zhang, Haifa Kassis, Rui Lan Zhang, Xiao Ming Zhang, Zheng Gang Zhang
ABSTRAKT
Stroke induces new myelinating oligodendrocytes that are involved in ischemic brain repair. Molecular mechanisms that regulate oligodendrogenesis have not been fully investigated. MicroRNAs (miRNAs) are small non-coding RNA molecules that post-transcriptionally regulate gene expression. MiR-146a has been reported to regulate immune response, but the role of miR-146a in oligodendrocyte progenitor cells (OPCs) remains unknown. Adult Wistar rats were subjected to the right middle cerebral artery occlusion (MCAo). In situ hybridization analysis with LNA probes against miR-146a revealed that stroke considerably increased miR-146a density in the corpus callosum and subventricular zone (SVZ) of the lateral ventricle of the ischemic hemisphere. In vitro, overexpression of miR-146a in neural progenitor cells (NPCs) significantly increased their differentiation into O4
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human IRAK1