Merck
NSE, a potential biomarker, is closely connected to diabetic peripheral neuropathy.
Diabetes care (2013-07-13)
Jianbo Li, Hongman Zhang, Min Xie, Lingfei Yan, Jiawei Chen, Hongxing Wang
ABSTRAKT
To explore the relationship between serum neuron-specific enolase (NSE) levels and diabetic neuropathy. Type 1 or 2 diabetic and healthy control subjects (n = 568) were randomly enrolled in a cross-sectional study. Diabetic neuropathy status was documented by the presence of clinical symptoms or signs, electromyography, quantitative sensory tests, and cardiac autonomic neuropathy tests. The severity of the neuropathy was staged by composite scores. Serum NSE was measured using electrochemiluminescence immunoassay. The demographic and clinical variables were obtained through an interviewer questionnaire. Serum NSE levels increased slightly in diabetic subjects compared with normal subjects (9.1 [1.5] vs. 8.7 [1.7], P = 0.037), and the levels increased greatly in diabetic subjects with neuropathy compared with those without (10.8 [2.8] vs. 9.1 [1.5], P = 0.000). The association of NSE with diabetic neuropathy was independent of the hyperglycemic state (fasting blood glucose, HbA1c, duration, and the type of diabetes) and other potential confounders affecting NSE levels (e.g., age, sex, and renal status) (odds ratio 1.48 [1.13-1.74], P = 0.001). In addition, NSE levels increased with and were closely correlated to the stages of neuropathy (r = 0.63 [0.52-0.74], P = 0.000). The optimal cutoff point for serum NSE levels to distinguish patients with diabetic neuropathy from those without was 10.10 μg/L, with a sensitivity of 66.3% and a specificity of 72.5%. Serum NSE levels are closely associated with peripheral neuropathy in patients with diabetes. Future studies are warranted to clarify the relationship.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Neuron-Specific Enolase human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE)