Merck
  • Diversification of echinomycin molecular structure by way of chemoenzymatic synthesis and heterologous expression of the engineered echinomycin biosynthetic pathway.
Diversification of echinomycin molecular structure by way of chemoenzymatic synthesis and heterologous expression of the engineered echinomycin biosynthetic pathway.
Current opinion in chemical biology (2009-03-13)
Kenji Watanabe, Hiroki Oguri, Hideaki Oikawa
ABSTRAKT
Echinomycin, a bis-intercalator antitumor cyclic peptide, is biosynthesized by a unique nonribosomal peptide synthetase (NRPS). Successful heterologous expression of the whole gene cluster of echinomycin in Escherichia coli let us to investigate a further application of echinomycin NRPS. To construct a cyclic peptide library, our approach through both chemoenzymatic and rational genetic engineering has been successfully demonstrated. These achievements provided the further support that E. coli-based system can serve as a flexible yet robust platform for producing complex natural products and their analogs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Echinomycin, ≥98% (HPLC)