Merck
Platelet-derived growth factor-D promotes fibrogenesis of cardiac fibroblasts.
American journal of physiology. Heart and circulatory physiology (2013-04-16)
Tieqiang Zhao, Wenyuan Zhao, Yuanjian Chen, Victoria S Li, Weixin Meng, Yao Sun
ABSTRAKT
Platelet-derived growth factor (PDGF)-D is a newly recognized member of the PDGF family with its role just now being understood. Our previous study shows that PDGF-D and its receptors (PDGFR-β) are significantly increased in the infarcted heart, where PDGFR-β is primarily expressed by fibroblasts, indicating the involvement of PDGF-D in the development of cardiac fibrosis. In continuing with these findings, the current study explored the molecular basis of PDGF-D on fibrogenesis. Rat cardiac fibroblasts were isolated and treated with PDGF-D (200 ng/ml medium). The potential regulation of PDGF-D on fibroblast growth, phenotype change, collagen turnover, and the transforming growth factor (TGF)-β pathway were explored. We found: 1) PDGF-D significantly elevated cardiac fibroblast proliferation, myofibroblast (myoFb) differentiation, and type I collagen secretion; 2) matrix metalloproteinase (MMP)-1, MMP-2, and MMP-9 protein levels were significantly elevated in PDGF-D-treated cells, which were coincident with increased expressions of tissue inhibitor of metalloproteinase (TIMP)-1 and TIMP-2; 3) PDGF-D significantly enhanced TGF-β1 synthesis, which was eliminated by TGF-β blockade with small-interfering RNA (siRNA); 4) the stimulatory role of PDGF-D on fibroblast proliferation and collagen synthesis was abolished by TGF-β blockade; and 5) TGF-β siRNA treatment significantly suppressed PDGF-D synthesis in fibroblasts. These observations indicate that PDGF-D promotes fibrogenesis through multiple mechanisms. Coelevations of TIMPs and MMPs counterbalance collagen degradation. The profibrogenic role of PDGF-D is mediated through activation of the TGF-β1 pathway. TGF-β1 exerts positive feedback on PDGF-D synthesis. These findings suggest the potential therapeutic effect of PDGFR blockade on interstitial fibrosis in the infarcted heart.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
QCM Chemotaxis Cell Migration Assay, 24-well (8 µm), colorimetric, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with colorimetric detection.