Merck
Math1 gene transfer based on the delivery system of quaternized chitosan/Na-carboxymethyl-beta-cyclodextrin nanoparticles.
Journal of nanoscience and nanotechnology (2010-12-09)
Li-Li Ren, Yan Wu, Dong Han, Li-Dong Zhao, Quan-Mei Sun, Wei-Wei Guo, Jian-He Sun, Nan Wu, Xing-Qi Li, Suo-Qiang Zhai, Dong-Yi Han, Wie-Yen Young, Shi-Ming Yang
ABSTRAKT
Mammalian cochlear hair cells don't regenerate naturally after injury, which usually leave permanent hearing loss. Math1 gene is a positive regulator of hair cell differentiation during cochlear development and was proved to be very critical in hair cell regeneration in deaf animals. Generating new cochlear hair cells by forced Math1 expression may be a cure for hearing loss. However, satisfying gene delivering vectors in gene therapy are not available. We combined quaternized chitosan (QCS) with Na-carboxymethyl-beta-cyclodextrin (CM-beta-CD) as novel non-viral vector, which adsorbs pRK5-Math1-EGFP perfectly at the mass ratio of 4:1. In vitro cell transfection can reach a 40% transfect efficiency and relatively low cytotoxity than liposomes. These results suggest that QCS/CM-beta-CD nanoparticle complexes could be a novel non-viral gene carrier in further clinical application.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carboxymethyl-β-cyclodextrin sodium salt