Merck
  • Synthesis of uridine 5'-(2-acetamido-2,4-dideoxy-4-fluoro-alpha-D-galactopyranosyl) diphosphate and uridine 5'-(2-acetamido-2,6-dideoxy-6-fluoro-alpha-D-glucopyranosyl) diphosphate.
Synthesis of uridine 5'-(2-acetamido-2,4-dideoxy-4-fluoro-alpha-D-galactopyranosyl) diphosphate and uridine 5'-(2-acetamido-2,6-dideoxy-6-fluoro-alpha-D-glucopyranosyl) diphosphate.
Carbohydrate research (1988-12-31)
R L Thomas, S A Abbas, K L Matta
ABSTRAKT
Benzyl 2-acetamido-3,6-di-O-benzyl-2-deoxy-alpha-D-glucopyranoside was converted into its 4-O-(methylsulfonyl) derivative (2) by treatment with methanesulfonyl chloride in pyridine. Displacement of the methylsulfonyloxy group of 2 with fluoride ion afforded benzyl 2-acetamido-3,6-di-O-benzyl-2,4-dideoxy-4-fluoro-alpha-D-galactopyranosi de, which on hydrogenolysis, followed by acetylation, furnished 2-acetamido-1,3,6-tri-O-acetyl-2,4-dideoxy-4-fluoro-D-galactopyranose. Treatment of this and of 2-acetamido-1,3,4-tri-O-acetyl-2,6-dideoxy-6-fluoro-D-glucopyranose with trimethylsilyl trifluoromethanesulfonate in 1,2-dichloroethane at approximately 50 degrees afforded the 4-deoxy-4-fluoro- or the 6-deoxy-6-fluoro-oxazolines (5) and (11), respectively. Reaction of 5 and 11 with dibenzyl phosphate in 1,2-dichloroethane produced the alpha-linked dibenzyl phosphate derivatives 6 and 12, respectively. Catalytic hydrogenation of 6 provided 2-acetamido-3,6-di-O-acetyl-2,4-dideoxy-4-fluoro-alpha-D-galactopyranosy l phosphate (7), and that of 12 gave 2-acetamido-3,4-di-O-acetyl-2,6-dideoxy-6-fluoro-alpha-D-glucopyranosyl phosphate (13). Coupling of 7 and 13 with uridine 5'-monophosphomorpholidate in dry pyridine at approximately 37 degrees, followed by O-deacetylation, furnished the title compounds, respectively, isolated and characterized as their respective dilithium salts.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Guanosine 5′-monophosphomorpholidate 4-morpholine-N,N′-dicyclohexylcarboxamidine salt, ≥90%