Merck
  • Glycerolipid Composition of the Red Macroalga Agarophyton Chilensis and Comparison to the Closely Related Agarophyton Vermiculophyllum Producing Different Types of Eicosanoids.
Glycerolipid Composition of the Red Macroalga Agarophyton Chilensis and Comparison to the Closely Related Agarophyton Vermiculophyllum Producing Different Types of Eicosanoids.
Marine drugs (2019-02-06)
Masaki Honda, Takashi Ishimaru, Yutaka Itabashi, Mikhail Vyssotski
ABSTRAKT
The red macroalga Agarophyton chilensis is a well-known producer of eicosanoids such as hydroxyeicosatetraenoic acids, but the alga produces almost no prostaglandins, unlike the closely related A. vermiculophyllum. This indicates that the related two algae would have different enzyme systems or substrate composition. To carry out more in-depth discussions on the metabolic pathway of eicosanoids between the two algae, we investigated the characteristics of glycerolipids, which are the substrates of eicosanoids production, of A. chilensis and compared them to the reported values of A. vermiculophyllum. In A. chilensis, monogalactosyldiacylglycerol (MGDG), digalactosyldiacylglycerol (DGDG), sulfoquinovosyldiacylglycerol (SQDG), and phosphatidylcholine (PC) were the major lipid classes and accounted for 44.4% of the total lipid extract. The predominant fatty acids were arachidonic acid (20:4n-6), an eicosanoids precursor, and palmitic acid (16:0). The 20:4n-6 content was extremely high in MGDG and PC (>70%), and the 16:0 content was extremely high in DGDG and SQDG (>40%). A chiral-phase HPLC analysis showed that fatty acids were esterified at the sn-1 and sn-2 positions of those lipids. The glycerolipid molecular species were determined by reversed-phase HPLC⁻ESI⁻MS analysis. The main glycerolipid molecular species were 20:4n-6/20:4n-6 (sn-1/sn-2) for MGDG (63.8%) and PC (48.2%), 20:4n-6/16:0 for DGDG (71.1%) and SQDG (29.4%). These lipid characteristics of A. chilensis were almost the same as those of A. vermiculophyllum. Hence, the differences of the eicosanoids producing ability between the two algae would not be due to the difference of substrate composition but the difference of enzyme system.