Wstecz
Poly(fluorescein isothiocyanate allylamine hydrochloride)
Products
Marka
Sigma-Aldrich (1)
Filtruj produkty
Showing 1-1 of 1
Porównaj
Numer produktu
Opis
Cennik
630217
Poly(fluorescein isothiocyanate allylamine hydrochloride),
Poly(allylamine hydrochloride) : Fluorescein isothiocyanate 50:1