57663
Synonim(y):
Upchurch PEEK Fingertight Union
Właściwości
0.020 in.
10-32 thread
Opis
Pokaż różnice
1 of 1
Cross
57659
Union
57658
57661
Tee
bore 0.020 in., 10-32 thread
bore 0.010 in., 10-32 thread
0.010 in.
Wybierz jedną z najnowszych wersji:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.