Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 695,00 zł |
Informacje o tej pozycji
695,00 zł
material
F217/PTFE cap liner, PTFE liner, clear glass vial, green melamine resin cap (solid-top)
feature
closure type green melamine solid-top cap, closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
21 mm × 70 mm
volume
15 mL
fitting
thread for 18-400
application(s)
general analytical
General description
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 15 mL, clear glass vial, O.D. × H 21 mm × 70 mm, thread for 18-400 | description volume 2 mL, clear glass vial (standard opening - 4.6 mm), O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425 | description volume 40 mL, amber glass vial, O.D. × H 29 mm × 82 mm, thread for 24-400 | description volume 22 mL, amber glass vial, O.D. × H 23 mm × 85 mm, thread for 20-400 |
| volume 15 mL | volume 2 mL | volume 40 mL | volume 22 mL |
| fitting thread for 18-400 | fitting thread for 8-425 | fitting thread for 24-400 | fitting thread for 20-400 |
| feature closure type screw top vial, closure type green melamine solid-top cap | feature closure type green melamine solid-top cap, closure type screw top vial | feature closure type screw top vial, closure type green melamine solid-top cap | feature closure type green melamine solid-top cap, closure type screw top vial |
| material PTFE liner, F217/PTFE cap liner, green melamine resin cap (solid-top), clear glass vial | material green melamine resin cap (solid-top), PTFE liner, F217/PTFE cap liner, clear glass vial (standard opening - 4.6 mm) | material green melamine resin cap (solid-top), F217/PTFE cap liner, PTFE liner, amber glass vial | material green melamine resin cap (solid-top), amber glass vial, F217/PTFE cap liner, PTFE liner |
| packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 100 ea |
| O.D. × H 21 mm × 70 mm | O.D. × H - | O.D. × H 29 mm × 82 mm | O.D. × H 23 mm × 85 mm |
Still not finding the right product?
Explore all of our products under Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?