Informacje o tej pozycji
UNSPSC Code:
41122001
NACRES:
NB.03
Pomoc techniczna
Pozwól nam pomócmaterial
bevel tip stainless steel needle (point style 2)
needle size
26 ga
description
replacement needles
packaging
pkg of 5 ea
manufacturer/tradename
SGE 037110
volume
10 μL
application(s)
general analytical
General description
RN syringes allow easy replacement of needles that get damaged in the field. The needle is seated at zero graduation mark.
Still not finding the right product?
Explore all of our products under SGE Needles for RN syringes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?