Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 182,00 zł |
Informacje o tej pozycji
182,00 zł
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
SWAGELOK® 400-7-4
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/4 in. SWAGELOK, 1/4 in. female NPT
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description SWAGELOK® 400-7-4, stainless steel, 1/4 in. SWAGELOK, 1/4 in. female NPT | description SWAGELOK® 400-7-2, stainless steel, 1/4 in. SWAGELOK, 1/8 in. female NPT | description brass, 1/8 in. SWAGELOK, 1/8 in. male NPT, 200-1-2 | description brass, 1/8 in. SWAGELOK, 1/4 in. male NPT, 200-1-4 |
| fitting 1/4 in. female NPT, 1/4 in. SWAGELOK | fitting 1/8 in. female NPT, 1/4 in. SWAGELOK | fitting 1/8 in. SWAGELOK, 1/8 in. male NPT | fitting 1/4 in. male NPT, 1/8 in. SWAGELOK |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 2 ea | packaging pkg of 2 ea |
| manufacturer/tradename SWAGELOK® 400-7-4 | manufacturer/tradename SWAGELOK® 400-7-2 | manufacturer/tradename 200-1-2 | manufacturer/tradename 200-1-4 |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable |
| material stainless steel | material stainless steel | material brass | material brass |
Still not finding the right product?
Explore all of our products under Swagelok® Connector to Female NPT
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?