Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 352,00 zł |
Informacje o tej pozycji
352,00 zł
packaging
pkg of 10 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
I.D.
1/4 in.
compatibility
1/4 in. O.D. Column
General description
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 10 ea | description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 50 ea | description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 50 ea | description I.D. 1/4 in., 1/4 in. O.D. Column, pkg of 10 ea |
| I.D. 1/4 in. | I.D. 1/4 in. | I.D. 1/4 in. | I.D. 1/4 in. |
| compatibility 1/4 in. O.D. Column | compatibility 1/4 in. O.D. Column | compatibility 1/4 in. O.D. Column | compatibility 1/4 in. O.D. Column |
| packaging pkg of 10 ea | packaging pkg of 50 ea | packaging pkg of 50 ea | packaging pkg of 10 ea |
| technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable | technique(s) gas chromatography (GC): suitable using :Suitable |
Still not finding the right product?
Explore all of our products under Supeltex M-4 Packed Column Ferrule, 1/4 in. Column O.D.
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?